Assay Details
Target Gene Details
Entrez Gene ID: | 224902 |
Gene Name: | scaffold attachment factor B2 |
Gene Aliases: |
AA389433, AI255170, mKIAA0138 |
Location: |
Chr.17:56560971-56584672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Safb2 | NM_001029979.2 | NP_001025150.2 | ||
AK147798.1 | BAE28146.1 | |||
AK161772.1 | ||||
AK172898.1 | BAD32176.1 | |||
BC050855.1 | AAH50855.1 | |||
CN723671.1 |
