Assay Details
Target Gene Details
Entrez Gene ID: | 106512 |
Gene Name: | G-protein signalling modulator 3 (AGS3-like, C. elegans) |
Gene Aliases: |
AA960287, AGS4 |
Location: |
Chr.17:34589806-34591754 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpsm3 | NM_134116.5 | NP_598877.1 | ||
| AK144833.1 | ||||
| AK146245.1 | BAE27009.1 | |||
| AK155617.1 | BAE33348.1 | |||
| AK155863.1 | BAE33467.1 | |||
| BC021942.1 | AAH21942.1 | |||
| BC080804.1 | AAH80804.1 | |||
| BC085186.1 |

Genomic Map