Assay Details
Target Gene Details
Entrez Gene ID: | 17685 |
Gene Name: | mutS homolog 2 (E. coli) |
Gene Aliases: |
AI788990 |
Location: |
Chr.17:87672557-87723713 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh2 | NM_008628.2 | 5 | 944 | NP_032654.1 |
| AK051960.1 | 5 | 941 | ||
| AK157842.1 | 5 | 957 | BAE34223.1 | |
| AK166179.1 | 5 | 930 | ||
| BC047117.1 | 5 | 952 | AAH47117.1 | |
| BC050897.1 | 5 | 927 | AAH50897.1 | |
| U21011.1 | 5 | 961 | AAA75027.1 | |
| X81143.1 | 5 | 944 | CAA57049.1 |
More Information
Set Membership: |
|

Genomic Map