Assay Details
Target Gene Details
Entrez Gene ID: | 66416 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (B14.5a) |
Gene Aliases: |
14.5kDa, 2400007M02Rik, CI-B14.5a |
Location: |
Chr.17:33824572-33838316 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa7 | NM_023202.4 | NP_075691.1 | ||
| NR_103514.1 | 1 | 79 | ||
| NR_103515.1 | ||||
| NR_103516.1 | ||||
| AK002476.1 | BAB22129.1 | |||
| AK003468.1 | BAB22805.1 | |||
| AK010299.1 | BAB26832.1 | |||
| BC052817.1 | AAH52817.1 | |||
| BC055698.1 | AAH55698.1 | |||
| BX518947.1 | ||||
| BY605952.1 | ||||
| KJ650204.1 |
More Information
Set Membership: |
|

Genomic Map