Assay Details
Target Gene Details
Entrez Gene ID: | 50873 |
Gene Name: | Parkinson disease (autosomal recessive, juvenile) 2, parkin |
Gene Aliases: |
PRKN |
Location: |
Chr.17:10840384-12063370 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Park2 | NM_016694.3 | 6 | 767 | NP_057903.1 |
| AB019558.1 | 6 | 738 | BAA82404.1 | |
| AF250293.1 | 6 | 767 | AAG13890.1 | |
| AF250294.1 | 6 | 770 | AAG13891.1 | |
| AF250295.1 | 6 | 770 | AAG13892.1 | |
| BC113204.1 | 6 | 750 | AAI13205.1 | |
| FJ555554.1 | 4 | 520 | ||
| FJ555555.1 | ||||
| FJ555556.1 | 5 | 500 | ||
| FJ555557.1 | ||||
| GU272056.1 |

Genomic Map