Assay Details
Target Gene Details
Entrez Gene ID: | 27226 |
Gene Name: | phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma) |
Gene Aliases: |
R75400 |
Location: |
Chr.17:43568082-43612202 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g7 | NM_013737.5 | NP_038765.2 | ||
| AK005158.1 | BAB23849.1 | |||
| AK005210.1 | ||||
| AK051454.1 | BAC34647.1 | |||
| AK155678.1 | BAE33386.1 | |||
| BC010726.1 | AAH10726.1 | |||
| U34277.1 | AAC52274.1 |
More Information
Set Membership: |
|

Genomic Map