Assay Details
Target Gene Details
Entrez Gene ID: | 224860 |
Gene Name: | phospholipase C-like 2 |
Gene Aliases: |
PLC-L2, PRIP-2, Plce2, mKIAA1092 |
Location: |
Chr.17:50508633-50688494 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcl2 | NM_013880.3 | 5 | 3205 | NP_038908.2 |
| AB033615.1 | 5 | 3333 | BAA89457.1 | |
| AK134985.1 | 4 | 1536 | ||
| AK154288.1 | 5 | 3161 | BAE32490.1 | |
| AK165548.1 | 5 | 3211 | ||
| BC027746.1 | 5 | 3267 | AAH27746.1 |
More Information
Set Membership: |
|

Genomic Map