Assay Details
Target Gene Details
Entrez Gene ID: | 213389 |
Gene Name: | PR domain containing 9 |
Gene Aliases: |
BC012016, Dsbc1, G1-419-29, Meisetz, Rcr1, repro7 |
Location: |
Chr.17:15543072-15564379 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prdm9 | BC012016.1 | AAH12016.1 | ||
BC049903.1 |
