Assay Details
Target Gene Details
Entrez Gene ID: | 224903 |
Gene Name: | scaffold attachment factor B |
Gene Aliases: |
3110021E02Rik, 5330423C17Rik, AU018122, D18386, E130307D12, HAP, HET, SAF-B1, SAFB1 |
Location: |
Chr.17:56584760-56606294 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Safb | NM_001163300.1 | NP_001156772.1 | ||
| AK087504.1 |
More Information
Set Membership: |
|

Genomic Map