Assay Details
Target Gene Details
Entrez Gene ID: | 17256 |
Gene Name: | male enhanced antigen 1 |
Gene Aliases: |
Mea-1 |
Location: |
Chr.17:46679138-46683126 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mea1 | NM_001277309.1 | NP_001264238.1 | ||
NM_001277310.1 | NP_001264239.1 | |||
NM_001277311.1 | NP_001264240.1 | |||
NM_001277312.1 | NP_001264241.1 | |||
NM_010787.2 | NP_034917.1 | |||
NR_102392.1 | ||||
AK005106.1 | BAB23822.1 | |||
AK005369.1 | BAB23978.1 | |||
AK008194.1 | BAB25525.1 | |||
AK008305.1 | BAB25590.1 | |||
AK010863.1 | BAB27231.1 | |||
AK010904.1 | BAB27257.1 | |||
AK010962.1 | BAB27294.1 | |||
AK011189.1 | BAB27454.1 | |||
AK012617.1 | BAB28358.1 | |||
BG085669.2 | ||||
BY707372.1 | ||||
CJ042312.1 | ||||
D17341.1 | BAA04159.1 | |||
L10401.1 | AAA39520.1 | |||
M27938.1 | AAA39519.1 |
