Assay Details
Target Gene Details
Entrez Gene ID: | 268973 |
Gene Name: | NLR family, CARD domain containing 4 |
Gene Aliases: |
9530011P19Rik, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, Card12, IPAF |
Location: |
Chr.17:74425716-74459143 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrc4 | NM_001033367.3 | NP_001028539.1 | ||
AK035300.1 | ||||
AK143864.1 | BAE25573.1 | |||
AK171624.1 | BAE42570.1 |
