Assay Details
Target Gene Details
Entrez Gene ID: | 57256 |
Gene Name: | protease, serine 21 |
Gene Aliases: |
1700023E12Rik, TESP5, mT4, testisin |
Location: |
Chr.17:23867873-23873113 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss21 | NM_020487.4 | 5 | 633 | NP_065233.2 |
| AB033822.1 | 5 | 619 | BAB64263.1 | |
| AB059415.1 | 5 | 631 | BAB61788.1 | |
| AF176209.2 | 5 | 633 | AAF64407.2 | |
| AK006271.1 | 5 | 605 | ||
| AY005145.1 | 5 | 610 | AAG02255.1 | |
| BC049588.1 | 5 | 647 | AAH49588.1 | |
| BC116755.1 | 5 | 631 | ||
| BC116757.1 | 5 | 631 |
More Information
Set Membership: |
|

Genomic Map