Assay Details
Target Gene Details
| Entrez Gene ID: | 70835 | 
| Gene Name: | protease, serine 22 | 
| Gene Aliases: | 4733401N09Rik, BSSP-4, SP001LA | 
| Location: | Chr.17:23993534-24000735 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 1 - Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Prss22 | NM_133731.2 | NP_598492.2 | ||
| AB010778.1 | BAB20262.1 | |||
| AK014645.1 | ||||
| BC055854.1 | AAH55854.1 | |||
| CD773447.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top