Assay Details
Target Gene Details
Entrez Gene ID: | 213171 |
Gene Name: | protease, serine 27 |
Gene Aliases: |
CAPH2, Mpn, marapsin |
Location: |
Chr.17:24038243-24048378 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss27 | NM_175440.4 | NP_780649.1 | ||
| AF542056.1 | AAO27572.1 | |||
| AK080281.1 | BAC37864.1 | |||
| BC117009.1 | ||||
| BC117011.1 |
More Information
Set Membership: |
|

Genomic Map