Assay Details
Target Gene Details
Entrez Gene ID: | 18519 |
Gene Name: | K(lysine) acetyltransferase 2B |
Gene Aliases: |
A930006P13Rik, AI461839, AW536563, Pcaf, p/CAF |
Location: |
Chr.17:53566853-53672721 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat2b | NM_001190846.1 | NP_001177775.1 | ||
NM_020005.4 | NP_064389.2 | |||
AF254442.1 | AAF70498.1 | |||
AK030070.1 | ||||
AK132486.1 | ||||
AK156290.1 | BAE33658.1 | |||
AK211252.1 | ||||
BC082581.1 | AAH82581.1 | |||
BC138195.1 | ||||
BC145896.1 |
