Assay Details
Target Gene Details
Entrez Gene ID: | 268977 |
Gene Name: | latent transforming growth factor beta binding protein 1 |
Gene Aliases: |
9430031G15Rik, 9830146M04, Ltbp-1, Tgfb, b2b1000Clo |
Location: |
Chr.17:75005529-75392967 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbp1 | NM_019919.3 | 6 | 1602 | NP_064303.2 |
| NM_206958.2 | 2 | 463 | NP_996841.1 | |
| AF022889.1 | 6 | 1567 | AAC33307.1 | |
| AF280604.1 | 2 | 347 | AAG53890.1 | |
| AK036047.1 | 2 | 466 | ||
| BC094612.1 | 2 | 412 | AAH94612.1 | |
| CV559283.1 |
More Information
Set Membership: |
|

Genomic Map