Assay Details
Target Gene Details
Entrez Gene ID: | 18519 |
Gene Name: | K(lysine) acetyltransferase 2B |
Gene Aliases: |
A930006P13Rik, AI461839, AW536563, Pcaf, p/CAF |
Location: |
Chr.17:53566853-53672721 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat2b | NM_001190846.1 | NP_001177775.1 | ||
| NM_020005.4 | NP_064389.2 | |||
| AF254442.1 | AAF70498.1 | |||
| AK030070.1 | ||||
| AK132486.1 | ||||
| AK156290.1 | BAE33658.1 | |||
| BC082581.1 | AAH82581.1 | |||
| BC138195.1 | ||||
| BC145896.1 |

Genomic Map