Assay Details
Target Gene Details
Entrez Gene ID: | 433091 |
Gene Name: | patatin-like phospholipase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28857965-28890308 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpla1 | NM_001034885.3 | 4 | 1062 | NP_001030057.1 |
| AK132521.1 | 4 | 1063 | BAE21216.1 | |
| BC139043.1 | 4 | 769 |
More Information
Set Membership: |
|

Genomic Map