Assay Details
Target Gene Details
Entrez Gene ID: | 268935 |
Gene Name: | signal peptide, CUB domain, EGF-like 3 |
Gene Aliases: |
CEGF3, D030038I21Rik |
Location: |
Chr.17:28142100-28174851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scube3 | NM_001004366.1 | NP_001004366.1 | ||
| AY639609.1 | AAU08348.1 | |||
| BC079849.1 | AAH79849.1 |
More Information
Set Membership: |
|

Genomic Map