Assay Details
Target Gene Details
Entrez Gene ID: | 193796 |
Gene Name: | lysine (K)-specific demethylase 4B |
Gene Aliases: |
4732474L06Rik, Jmjd2b, mKIAA0876 |
Location: |
Chr.17:56326033-56403769 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm4b | NM_172132.2 | 1 | 61 | NP_742144.1 |
| AK078074.1 | 1 | 54 | ||
| AK129233.1 | BAC98043.1 | |||
| AK141879.1 | 1 | 38 | BAE24866.1 | |
| AK145151.1 | ||||
| BC007145.1 | AAH07145.1 | |||
| CA324771.1 | 1 | 61 | ||
| CJ093438.1 | 1 | 32 |
More Information
Set Membership: |
|

Genomic Map