Assay Details
Target Gene Details
Entrez Gene ID: | 116871 |
Gene Name: | metastasis associated 3 |
Gene Aliases: |
1110002J22Rik, mKIAA1266 |
Location: |
Chr.17:83706163-83823715 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mta3 | NM_001171052.1 | 16 | 1735 | NP_001164523.1 |
| NM_054082.2 | 16 | 1732 | NP_473423.1 | |
| AF288138.1 | 16 | 1686 | AAK83045.1 | |
| AK146721.1 | 16 | 1688 | BAE27384.1 | |
| AK146905.1 | 16 | 1688 | BAE27518.1 | |
| AK158866.1 | 15 | 1598 | BAE34701.1 |
More Information
Set Membership: |
|

Genomic Map