Assay Details
Target Gene Details
Entrez Gene ID: | 67676 |
Gene Name: | ribonuclease P 21 subunit |
Gene Aliases: |
0610037N01Rik |
Location: |
Chr.17:36255673-36257846 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpp21 | NM_026308.2 | 2 | 123 | NP_080584.1 |
| AJ504717.1 | 2 | 133 | CAD44293.1 | |
| AK002780.1 | 2 | 124 | BAB22353.1 | |
| AK003455.1 | 2 | 121 | BAB22800.1 | |
| AY251016.1 | 2 | 145 | AAP03887.1 | |
| AY251017.1 | AAP03888.1 | |||
| BC028491.1 | 2 | 133 | AAH28491.1 |

Genomic Map