Assay Details
Target Gene Details
Entrez Gene ID: | 15482 |
Gene Name: | heat shock protein 1-like |
Gene Aliases: |
Hsc70t, Msh5 |
Location: |
Chr.17:34972703-34979228 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hspa1l | NM_013558.2 | 1 | 124 | NP_038586.2 |
BC129894.1 | ||||
BC129895.1 | ||||
BY099756.1 | 1 | 126 | ||
D85732.1 | 1 | 95 | BAA32522.1 |
