Assay Details
Target Gene Details
Entrez Gene ID: | 116871 |
Gene Name: | metastasis associated 3 |
Gene Aliases: |
1110002J22Rik, mKIAA1266 |
Location: |
Chr.17:83706163-83823715 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mta3 | NM_001171052.1 | NP_001164523.1 | ||
| NM_001171053.1 | NP_001164524.1 | |||
| NM_001171054.1 | NP_001164525.1 | |||
| NM_054082.2 | NP_473423.1 | |||
| AF288138.1 | AAK83045.1 | |||
| AK129318.1 | BAC98128.1 | |||
| AK145942.1 | BAE26772.1 | |||
| AK146721.1 | BAE27384.1 | |||
| AK146905.1 | BAE27518.1 | |||
| AK147197.1 | BAE27755.1 | |||
| AK154856.1 | BAE32881.1 | |||
| AK158866.1 | BAE34701.1 | |||
| BC022124.1 | AAH22124.1 | |||
| BC099960.1 | ||||
| CA751287.1 |
More Information
Set Membership: |
|

Genomic Map