Assay Details
Target Gene Details
Entrez Gene ID: | 19015 |
Gene Name: | peroxisome proliferator activator receptor delta |
Gene Aliases: |
NUC-1, NUC1, Nr1c2, PPAR-beta, PPAR-delta, PPAR[b], PPARdelta, Pparb, Pparb/d |
Location: |
Chr.17:28232754-28301469 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppard | NM_011145.3 | 2 | 152 | NP_035275.1 |
| AK089913.1 | 2 | 100 | ||
| AK154567.1 | 2 | 142 | ||
| BC070398.1 | 2 | 103 | AAH70398.1 | |
| CA978861.1 | 2 | 152 |
More Information
Set Membership: |
|

Genomic Map