Assay Details
Target Gene Details
Entrez Gene ID: | 100043332 |
Gene Name: | ankyrin repeat domain 66 |
Gene Aliases: |
AU041848, D730048J04Rik, Gm4368 |
Location: |
Chr.17:43534173-43570729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd66 | NM_001254953.1 | NP_001241882.1 |
