Assay Details
Target Gene Details
Entrez Gene ID: | 71893 |
Gene Name: | NADPH oxidase organizer 1 |
Gene Aliases: |
2310034C04Rik, P41NOX, P41NOXA, P41NOXB, P41NOXC, Snx28, hslt |
Location: |
Chr.17:24696234-24700530 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Noxo1 | NM_027988.4 | 5 | 1366 | NP_082264.2 |
AF399754.1 | 4 | 352 | AAK94017.1 | |
AF539797.1 | 4 | 352 | AAN75142.1 | |
AK009605.1 | 4 | 461 | BAB26387.1 | |
AK088226.1 | 5 | 1325 | BAC40222.1 | |
AK157993.1 | 5 | 1367 | BAE34305.1 | |
BC019525.1 | 4 | 378 | AAH19525.1 | |
BC119527.1 | 4 | 454 | ||
BC119528.1 | 3 | 537 | ||
BC127235.1 | 4 | 454 | ||
BC127236.1 | 4 | 454 |
