Assay Details
Target Gene Details
Entrez Gene ID: | 74116 |
Gene Name: | peptidase inhibitor 16 |
Gene Aliases: |
1200009H11Rik, Cripi, PI-16 |
Location: |
Chr.17:29318796-29328903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pi16 | NM_023734.3 | NP_076223.3 | ||
| AK004672.1 | 4 | 2126 | ||
| AK157854.1 | 4 | 2438 | ||
| AK158339.1 | 4 | 2433 | ||
| BC039124.1 | AAH39124.1 | |||
| BC118026.1 |
More Information
Set Membership: |
|

Genomic Map