Assay Details
Target Gene Details
Entrez Gene ID: | 224829 |
Gene Name: | transcriptional regulating factor 1 |
Gene Aliases: |
9430096I18Rik, AI429294, B830015H24, RAPA, Trep-132, Trep132 |
Location: |
Chr.17:47140942-47359458 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trerf1 | NM_001097623.1 | NP_001091092.1 | ||
NM_172622.2 | NP_766210.1 | |||
AK046822.1 | BAC32884.1 | |||
AY081774.1 | AAN02288.1 | |||
BC051645.1 | AAH51645.1 | |||
BC059215.1 | AAH59215.1 | |||
BC108416.1 | AAI08417.1 |
