Assay Details
Target Gene Details
Entrez Gene ID: | 75956 |
Gene Name: | serine/arginine repetitive matrix 2 |
Gene Aliases: |
5033413A03Rik, AA410130, SRm300, mKIAA0324 |
Location: |
Chr.17:23790857-23824741 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrm2 | NM_175229.3 | NP_780438.2 | ||
| AI463539.1 | ||||
| AK017163.1 | ||||
| AK090123.1 | BAC41104.1 | |||
| AK122248.1 | BAC65530.1 | |||
| BC027781.1 | AAH27781.1 |
Target Gene Details
Entrez Gene ID: | 67673 |
Gene Name: | transcription elongation factor B (SIII), polypeptide 2 |
Gene Aliases: |
0610040H15Rik |
Location: |
Chr.17:23824740-23829109 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tceb2 | NM_026305.2 | NP_080581.1 | ||
| AK002223.1 | BAB21946.1 | |||
| AK002868.1 | BAB22417.1 | |||
| AK003277.1 | BAB22684.1 | |||
| AK008477.1 | BAB25690.1 | |||
| AK008534.2 | BAB25727.1 | |||
| AK012254.1 | BAB28121.1 | |||
| AK019181.1 | BAB31590.1 | |||
| AK019218.1 | BAB31608.1 | |||
| AK019358.1 | BAB31677.1 | |||
| AK152950.1 | ||||
| BC051927.1 | AAH51927.2 | |||
| BC056983.1 | AAH56983.1 |
More Information
Set Membership: |
|

Genomic Map