Assay Details
Target Gene Details
Entrez Gene ID: | 21425 |
Gene Name: | transcription factor EB |
Gene Aliases: |
Tcfeb, bHLHe35 |
Location: |
Chr.17:47737037-47792416 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfeb | NM_001161722.1 | NP_001155194.1 | ||
NM_001161723.1 | NP_001155195.1 | |||
NM_011549.3 | NP_035679.3 | |||
AF079095.1 | AAD20979.1 | |||
AK078755.1 | BAC37377.1 | |||
AK146016.1 | BAE26834.1 | |||
AK154971.1 | BAE32963.1 | |||
AK198293.1 | ||||
AK199475.1 | ||||
AK208283.1 | ||||
BC064789.1 | AAH64789.1 |
