Assay Details
Target Gene Details
Entrez Gene ID: | 24001 |
Gene Name: | T cell lymphoma invasion and metastasis 2 |
Gene Aliases: |
3000002F19Rik, STEF, mKIAA2016 |
Location: |
Chr.17:3326300-3519397 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tiam2 | NM_001122998.1 | NP_001116470.1 | ||
NM_011878.2 | NP_036008.2 | |||
AB022915.1 | BAA81823.1 | |||
AK129474.1 | BAC98284.1 | |||
BC067048.1 | AAH67048.1 | |||
BC079600.1 | AAH79600.1 |
