Assay Details
Target Gene Details
Entrez Gene ID: | 71367 |
Gene Name: | carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 |
Gene Aliases: |
5430438D01Rik |
Location: |
Chr.18:15451773-15760415 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chst9 | NM_199055.2 | NP_951010.1 | ||
| AB106879.1 | BAC87754.1 | |||
| AK017407.1 | ||||
| BC107361.1 | AAI07362.1 |
More Information
Set Membership: |
|

Genomic Map