Assay Details
Target Gene Details
Entrez Gene ID: | 328795 |
Gene Name: | ubiquitin associated and SH3 domain containing, A |
Gene Aliases: |
5830413C03Rik, C330001M22, Sts-2, TULA, TULA-1 |
Location: |
Chr.17:31207713-31253419 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubash3a | NM_177823.4 | 7 | 1046 | NP_808491.2 |
AK040572.1 | 7 | 1007 | ||
AK041690.1 | 7 | 1013 | BAE20600.1 | |
AK049106.1 | 6 | 946 | BAC33544.1 | |
AK169696.1 | 7 | 1001 | BAE41311.1 |
