Assay Details
Target Gene Details
Entrez Gene ID: | 24001 |
Gene Name: | T cell lymphoma invasion and metastasis 2 |
Gene Aliases: |
3000002F19Rik, STEF, mKIAA2016 |
Location: |
Chr.17:3326300-3519397 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tiam2 | NM_001122998.1 | 6 | 2238 | NP_001116470.1 |
NM_011878.2 | 6 | 2222 | NP_036008.2 | |
AB022915.1 | 6 | 2223 | BAA81823.1 | |
AK129474.1 | 6 | 2141 | BAC98284.1 | |
AK158137.1 | 6 | 2201 | BAE34377.1 | |
BC067048.1 | 6 | 2238 | AAH67048.1 | |
BC079600.1 | 6 | 2127 | AAH79600.1 |
