Assay Details
Target Gene Details
Entrez Gene ID: | 211961 |
Gene Name: | additional sex combs like 3 (Drosophila) |
Gene Aliases: |
C230079D11Rik, D430002O22Rik, D930044O18Rik, Gm329, Gm945 |
Location: |
Chr.18:22344077-22530227 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asxl3 | NM_001167777.1 | NP_001161249.1 | ||
| AK082659.1 | BAC38564.2 |
More Information
Set Membership: |
|

Genomic Map