Assay Details
Target Gene Details
Entrez Gene ID: | 63955 |
Gene Name: | CDK5 and Abl enzyme substrate 1 |
Gene Aliases: |
Cables, ik3-1 |
Location: |
Chr.18:11839274-11945627 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cables1 | NM_001146287.1 | NP_001139759.1 | ||
NM_022021.2 | NP_071304.2 | |||
AF133208.1 | AAG33933.1 | |||
AF328140.1 | AAG42916.1 | |||
AK156273.1 | BAE33652.1 | |||
AK158396.1 | BAE34486.1 | |||
BC145142.1 | ||||
BC150714.1 | ||||
BQ917292.1 | ||||
CX202925.1 |
