Assay Details
Target Gene Details
Entrez Gene ID: | 52538 |
Gene Name: | acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase) |
Gene Aliases: |
0610011L04Rik, AI255831, AI265397, D18Ertd240e |
Location: |
Chr.18:74779212-74806207 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaa2 | NM_177470.3 | 4 | 396 | NP_803421.1 |
| AK002555.1 | 4 | 386 | ||
| AK050101.1 | 4 | 398 | BAC34067.1 | |
| AK146007.1 | 4 | 411 | BAE26828.1 | |
| AK167567.1 | 4 | 397 | BAE39630.1 | |
| AK167715.1 | 4 | 394 | BAE39757.1 | |
| AK169359.1 | 4 | 397 | BAE41108.1 | |
| BC028901.1 | 4 | 391 | AAH28901.1 |
More Information
Set Membership: |
|

Genomic Map