Assay Details
Target Gene Details
Entrez Gene ID: | 74355 |
Gene Name: | SMC hinge domain containing 1 |
Gene Aliases: |
4931400A14Rik, AW554188, MommeD1, mKIAA0650 |
Location: |
Chr.17:71344489-71475343 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smchd1 | NM_028887.3 | 27 | 3656 | NP_083163.3 |
| AK016419.1 | 2 | 133 | BAB30222.1 | |
| AK129181.1 | 5 | 653 | BAC97991.1 | |
| BC058618.1 | 3 | 308 | AAH58618.1 | |
| BC062946.1 | 13 | 1501 | AAH62946.1 |

Genomic Map