Assay Details
Target Gene Details
Entrez Gene ID: | 21454 |
Gene Name: | t-complex protein 1 |
Gene Aliases: |
AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63 |
Location: |
Chr.17:12916329-12925067 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcp1 | NM_001290712.1 | 9 | 1292 | NP_001277641.1 |
NM_013686.4 | 10 | 1378 | NP_038714.2 | |
AK149611.1 | 10 | 1243 | BAE28989.1 | |
AK149755.1 | 10 | 1244 | BAE29063.1 | |
AK150666.1 | 8 | 2803 | ||
AK150744.1 | 8 | 2803 | ||
AK151068.1 | 10 | 1243 | BAE30084.1 | |
AK151445.1 | 9 | 1182 | BAE30407.1 | |
AK151903.1 | 8 | 2803 | ||
AK152633.1 | 8 | 2803 | ||
AK152641.1 | 10 | 1235 | BAE31381.1 | |
AK152759.1 | 10 | 1243 | ||
AK153037.1 | 10 | 1243 | ||
AK153430.1 | 9 | 1182 | BAE31988.1 | |
AK165665.1 | 10 | 1245 | BAE38327.1 | |
AK166828.1 | 10 | 1242 | BAE39052.1 | |
AK166966.1 | 10 | 1266 | BAE39149.1 | |
AK167529.1 | 10 | 1243 | BAE39599.1 | |
AK178327.1 | 2 | 100 | ||
AK199928.1 | 2 | 99 | ||
BC003809.1 | 10 | 1226 | AAH03809.1 | |
D90344.1 | 10 | 1193 | BAA14356.1 | |
M12899.1 | 10 | 1201 | AAA40338.1 | |
M20130.1 | 3 | 266 | AAA40337.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
