Assay Details
Target Gene Details
Entrez Gene ID: | 11946 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1 |
Gene Aliases: |
AI035633, AL022851, AL023067, Atpm, D18Ertd206e, Mom2 |
Location: |
Chr.18:77773768-77782869 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
More Information
Set Membership: |
|

Genomic Map