Assay Details
Target Gene Details
Entrez Gene ID: | 74355 |
Gene Name: | SMC hinge domain containing 1 |
Gene Aliases: |
4931400A14Rik, AW554188, MommeD1, mKIAA0650 |
Location: |
Chr.17:71344489-71475343 on Build GRCm38 |
Assay Gene Location: | Within Exon 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smchd1 | NM_028887.3 | 45 | 5779 | NP_083163.3 |
| AK016419.1 | 20 | 2256 | BAB30222.1 | |
| AK030247.1 | 13 | 1361 | ||
| AK129181.1 | 23 | 2776 | BAC97991.1 | |
| BC044905.1 | 9 | 926 | AAH44905.1 | |
| BC058205.1 | 11 | 1239 | AAH58205.1 | |
| BC058618.1 | 21 | 2431 | AAH58618.1 | |
| BC062946.1 | 31 | 3624 | AAH62946.1 |

Genomic Map