Assay Details
Target Gene Details
Entrez Gene ID: | 21454 |
Gene Name: | t-complex protein 1 |
Gene Aliases: |
AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63 |
Location: |
Chr.17:12916329-12925067 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcp1 | NM_001290712.1 | NP_001277641.1 | ||
| NM_013686.4 | NP_038714.2 | |||
| AK149611.1 | BAE28989.1 | |||
| AK149755.1 | BAE29063.1 | |||
| AK150666.1 | ||||
| AK150744.1 | ||||
| AK151068.1 | BAE30084.1 | |||
| AK151445.1 | BAE30407.1 | |||
| AK151903.1 | ||||
| AK152633.1 | ||||
| AK152641.1 | BAE31381.1 | |||
| AK152759.1 | ||||
| AK153037.1 | ||||
| AK153430.1 | BAE31988.1 | |||
| AK165665.1 | BAE38327.1 | |||
| AK166828.1 | BAE39052.1 | |||
| AK166966.1 | BAE39149.1 | |||
| AK167529.1 | BAE39599.1 | |||
| BC003809.1 | AAH03809.1 | |||
| D90344.1 | BAA14356.1 | |||
| M12899.1 | AAA40338.1 |
More Information
Set Membership: |
|

Genomic Map