Assay Details
Target Gene Details
Entrez Gene ID: | 12322 |
Gene Name: | calcium/calmodulin-dependent protein kinase II alpha |
Gene Aliases: |
CaMKII, R74975, mKIAA0968 |
Location: |
Chr.18:60925580-60988152 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2a | NM_001286809.1 | NP_001273738.1 | ||
| NM_009792.3 | NP_033922.1 | |||
| NM_177407.4 | NP_803126.1 | |||
| AK083245.1 | BAC38829.1 | |||
| AK122410.2 | BAC65692.3 | |||
| BB659027.1 | ||||
| BC031745.1 | AAH31745.1 | |||
| X14836.1 | CAA32946.1 | |||
| X87142.1 | CAA60620.1 |
More Information
Set Membership: |
|

Genomic Map