Assay Details
Target Gene Details
Entrez Gene ID: | 12322 |
Gene Name: | calcium/calmodulin-dependent protein kinase II alpha |
Gene Aliases: |
CaMKII, R74975, mKIAA0968 |
Location: |
Chr.18:60925580-60988152 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2a | NM_177407.4 | 2 | 248 | NP_803126.1 |
| AK083245.1 | 2 | 249 | BAC38829.1 | |
| AK122410.2 | 2 | 244 | BAC65692.3 | |
| AK163179.1 | 2 | 249 | ||
| BC031745.1 | 2 | 229 | AAH31745.1 | |
| X14836.1 | 2 | 232 | CAA32946.1 |
More Information
Set Membership: |
|

Genomic Map