Assay Details
Target Gene Details
Entrez Gene ID: | 210510 |
Gene Name: | tudor domain containing 6 |
Gene Aliases: |
AB097085, Tdr2 |
Location: |
Chr.17:43615334-43631350 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdrd6 | NM_001161366.1 | NP_001154838.1 | ||
| NM_001161367.1 | NP_001154839.1 | |||
| NM_198418.2 | NP_940810.2 | |||
| AB097085.1 | BAD01486.1 | |||
| BC138353.1 | ||||
| BC145248.1 |
More Information
Set Membership: |
|

Genomic Map