Assay Details
Target Gene Details
Entrez Gene ID: | 77106 |
Gene Name: | transmembrane protein 181A |
Gene Aliases: |
5930418K15Rik, C76977, Gpr178, Tmem181, mKIAA1423 |
Location: |
Chr.17:6256865-6308314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem181a | NM_001033178.3 | NP_001028350.1 | ||
| AK122507.1 | BAC65789.3 | |||
| AK154554.1 | BAE32673.1 | |||
| AK178082.1 | ||||
| AK182609.1 | ||||
| AK184853.1 | ||||
| BQ832419.1 |

Genomic Map