Assay Details
Target Gene Details
Entrez Gene ID: | 224807 |
Gene Name: | transmembrane protein 63b |
Gene Aliases: |
BC026370 |
Location: |
Chr.17:45660172-45686254 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem63b | NM_198167.3 | NP_937810.2 | ||
AK090200.1 | BAC41131.1 | |||
AK159673.1 | BAE35277.1 | |||
BC059283.1 | AAH59283.1 |
