Assay Details
Target Gene Details
Entrez Gene ID: | 66530 |
Gene Name: | UBX domain protein 6 |
Gene Aliases: |
1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2 |
Location: |
Chr.17:56068249-56074989 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubxn6 | NM_024432.2 | NP_077752.1 | ||
AF272895.1 | AAK13259.1 | |||
AK008942.1 | ||||
AK009117.1 | BAB26082.1 | |||
AK075705.1 | ||||
AK161273.1 | BAE36284.1 | |||
AK205258.1 | ||||
BC016412.1 | AAH16412.1 | |||
BC049963.1 | AAH49963.1 |
