Assay Details
Target Gene Details
Entrez Gene ID: | 64074 |
Gene Name: | SPARC related modular calcium binding 2 |
Gene Aliases: |
1700056C05Rik, 5430426J21Rik, Smoc2l |
Location: |
Chr.17:14279487-14404790 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smoc2 | NM_022315.2 | NP_071710.2 | ||
| AJ249901.1 | CAC10355.1 | |||
| AK030958.1 | BAC27193.1 | |||
| AK159749.1 | BAE35341.1 | |||
| AK160002.1 | BAE35550.1 | |||
| BC019527.1 | AAH19527.1 |
More Information
Set Membership: |
|

Genomic Map